Decyltriethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Triethoxy(decyl)silane

SILS05600.10 SILS05600.25 SILS05600.100
SILNSILS05600.10EA 0 EUR
SILNSILS05600.10 SILNSILS05600.25 SILNSILS05600.100
Decyltriethoxysilan
Decyltriethoxysilan
Formel: C₁₆H₃₆O₃Si
Molecular Weight: 304.55 g/mol
MDL: MFCD00069132
CAS-Nummer: 2943-73-9
EINECS: 220-940-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR