Decyltrimethoxysilan
Lieferant: CHEMPUR SILANE
SILS05625.10
SILS05625.25
SILS05625.100
SILNSILS05625.10EA
0
EUR
SILNSILS05625.10
SILNSILS05625.25
SILNSILS05625.100
Decyltrimethoxysilan
Decyltrimethoxysilan
Formel:
C₁₃H₃₀O₃Si Molecular Weight: 262.46 g/mol |
MDL:
MFCD00069135 CAS-Nummer: 5575-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...