Ethyltriethoxysilan 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Triethoxy(ethyl)silan

L16495.AC L16495.AE
L16495.ACEA 45.5 EUR
L16495.AC L16495.AE
Ethyltriethoxysilan 96%
Ethyltriethoxysilan

Purity: 96% CAS No.: 78-07-9 Molecular Formula: (CH3CH2O)2SiCH2CH3 Formula Weight: 192.33 Flash Point: 40°(104°F) Refractive Index: 1.3920 Melting Point: -78° Boiling Point: 158-159° Beilstein Reference: 1362028 UN No.: 1993 MDL No.: MFCD00026754

Formel: C₈H₂₀O₃Si
Molecular Weight: 192,33 g/mol
Siedepunkt: 158…159 °C
Schmelzpunkt: –78 °C
Dichte: 0,896
Flammpunkt: 40 °C (104 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00026754
CAS-Nummer: 78-07-9
EINECS: 201-080-1
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR