Ethyltriethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Triethoxy(ethyl)silan

SILS08850.25 SILS08850.100
SILNSILS08850.25EA 0 EUR
SILNSILS08850.25 SILNSILS08850.100
Ethyltriethoxysilan
Ethyltriethoxysilan
Formel: C₈H₂₀O₃Si
Molecular Weight: 192,33 g/mol
Siedepunkt: 158 °C (1013 hPa)
Dichte: 0,9207 g/cm³ (0 °C)
MDL: MFCD00026754
CAS-Nummer: 78-07-9
EINECS: 201-080-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR