Ethyltrimethoxysilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trimethoxy(ethyl)silan
SILS08875.10
SILS08875.25
SILNSILS08875.10EA
0
EUR
SILNSILS08875.10
SILNSILS08875.25
Ethyltrimethoxysilan
Ethyltrimethoxysilan
Formel:
C₅H₁₄O₃Si Molecular Weight: 150.25 g/mol Siedepunkt: 123…124 °C (1013 hPa) Dichte: 0.94 g/cm³ (25 °C) |
MDL:
MFCD00051541 CAS-Nummer: 5314-55-6 EINECS: 226-172-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...