Hexyltriethoxysilan ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Triethoxy(hexyl)silane
L16521.14
L16521.22
L16521.14EA
43.1
EUR
L16521.14
L16521.22
Hexyltriethoxysilan ≥97%
Hexyltriethoxysilan
Purity: 97%
CAS No.: 18166-37-5
Molecular Formula: C12H28O3Si
Formula Weight: 248.44
Flash Point: 95°(203°F)
Boiling Point: 115°/18mm
MDL No.: MFCD00087744
Formel:
C₁₂H₂₈O₃Si Molecular Weight: 248,44 g/mol Siedepunkt: 115 °C (18 mmHg) Dichte: 0,860 Flammpunkt: 95 °C (203 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00087744 CAS-Nummer: 18166-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...