Hexyltriethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Triethoxy(hexyl)silane

SILS09950.25 SILS09950.100
SILNSILS09950.25EA 0 EUR
SILNSILS09950.25 SILNSILS09950.100
Hexyltriethoxysilan
Hexyltriethoxysilan
Formel: C₁₂H₂₈O₃Si
Molecular Weight: 248,44 g/mol
MDL: MFCD00087744
CAS-Nummer: 18166-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR