Methyltriethoxysilan ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L04059.AK L04059.AU L04059.PG
L04059.AKEA 48.4 EUR
L04059.AK L04059.AU L04059.PG
Methyltriethoxysilan ≥98%
Methyltriethoxysilan

Purity: 98% CAS No.: 2031-67-6 Molecular Formula: C7H18O3Si Formula Weight: 178.30 Flash Point: 23°(73°F) Refractive Index: 1.3835 Odour: Mild Melting Point: -50° Boiling Point: 141-143° Beilstein Reference: 1742453 UN No.: 1993 MDL No.: MFCD00009066

Formel: C₇H₁₈O₃Si
Siedepunkt: 141…143 °C
Schmelzpunkt: < –50 °C
Dichte: 0,897
Flammpunkt: 23 °C (73 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009066
CAS-Nummer: 2031-67-6
EINECS: 217-983-9
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR