Methyltrimethoxysilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trimethoxy(methyl)silan
SILS11825.25
SILS11825.100
SILNSILS11825.25EA
0
EUR
SILNSILS11825.25
SILNSILS11825.100
Methyltrimethoxysilan
Methyltrimethoxysilan
Formel:
C₄H₁₂O₃Si Molecular Weight: 136.22 g/mol Siedepunkt: 102 Schmelzpunkt: 11 Dichte: 0.955 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008342 CAS-Nummer: 1185-55-3 EINECS: 214-685-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...