Octadecyltriethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Triethoxy(octadecyl)silane

SILS12276.10 SILS12276.25 SILS12276.100
SILNSILS12276.10EA 0 EUR
SILNSILS12276.10 SILNSILS12276.25 SILNSILS12276.100
Octadecyltriethoxysilan
Octadecyltriethoxysilan
Formel: C₂₄H₅₂O₃Si
Molecular Weight: 416.76 g/mol
MDL: MFCD00039875
CAS-Nummer: 7399-00-0
EINECS: 230-995-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR