Octadecyltrimethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Trimethoxy(octadecyl)silan

SILS12325.25 SILS12325.100
SILNSILS12325.25EA 0 EUR
SILNSILS12325.25 SILNSILS12325.100
Octadecyltrimethoxysilan
Octadecyltrimethoxysilan
Formel: C₂₁H₄₆O₃Si
Molecular Weight: 374.68 g/mol
MDL: MFCD00043060
CAS-Nummer: 3069-42-9
EINECS: 221-339-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR