Propyltriethoxysilan ≥97%
Lieferant: Thermo Scientific
B20997.22
B20997.36
B20997.22EA
107
EUR
B20997.22
B20997.36
Propyltriethoxysilan ≥97%
Propyltriethoxysilan
Purity: 97%
CAS No.: 2550-02-9
Molecular Formula: CH3(CH2)2Si(OCH2CH3)3
Formula Weight: 206.36
Flash Point: 57°(134°F)
Refractive Index: 1.3965
Boiling Point: 179-180°
Beilstein Reference: 1748158
UN No.: 1993
MDL No.: MFCD00026756
Formel:
C₉H₂₂O₃Si Molecular Weight: 206,36 g/mol |
MDL:
MFCD00026756 CAS-Nummer: 2550-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...