Propyltriethoxysilan

Lieferant: CHEMPUR SILANE

SILS14100.25 SILS14100.100
SILNSILS14100.25EA 0 EUR
SILNSILS14100.25 SILNSILS14100.100
Propyltriethoxysilan
Propyltriethoxysilan
Formel: C₉H₂₂O₃Si
Molecular Weight: 206,36 g/mol
MDL: MFCD00026756
CAS-Nummer: 2550-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR