Propyltrimethoxysilan ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Trimethoxy(propyl)silan
B21033.14
B21033.22
B21033.36
B21033.14EA
24.2
EUR
B21033.14
B21033.22
B21033.36
Propyltrimethoxysilan ≥98%
Propyltrimethoxysilan
Purity: 98+%
CAS No.: 1067-25-0
Molecular Formula: CH3(CH2)2Si(OCH3)3
Formula Weight: 164.28
Flash Point: 34°(93°F)
Refractive Index: 1.3900
Boiling Point: 142-143°
Beilstein Reference: 2070458
UN No.: 1993
MDL No.: MFCD00043026
Formel:
C₆H₁₆O₃Si Molecular Weight: 164,28 g/mol Siedepunkt: 142…143 °C Dichte: 0,935 Flammpunkt: 34 °C (93 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043026 CAS-Nummer: 1067-25-0 EINECS: 213-926-7 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...