(3-Mercaptopropyl)triethoxysilan ≥94%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Triethoxysilyl)-1-propanthiol, 3-(Triethoxysilyl)propan-1-thiol, Triethoxy(3-mercaptopropyl)silan
B21191.14
B21191.22
B21191.14EA
81.9
EUR
B21191.14
B21191.22
(3-Mercaptopropyl)triethoxysilan ≥94%
(3-Mercaptopropyl)triethoxysilan
Purity: 94%
CAS No.: 14814-09-6
Molecular Formula: C9H22O3SSi
Formula Weight: 238.42
Flash Point: 88°(190°F)
Physical Form: Liquid
Density: 0.98
Refractive Index: 1.4370
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless
Odour: Unpleasant
Boiling Point: 210°
Beilstein Reference: 2039575
MDL No.: MFCD00053924
Formel:
HSH₂CCH₂CH₂Si(OCH₂CH₃)₃ Molecular Weight: 238,42 g/mol Siedepunkt: 210 °C Dichte: 0,98 Flammpunkt: 88 °C (190 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053924 CAS-Nummer: 14814-09-6 EINECS: 238-883-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...