(3-Mercaptopropyl)triethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: 3-(Triethoxysilyl)-1-propanthiol, 3-(Triethoxysilyl)propan-1-thiol, Triethoxy(3-mercaptopropyl)silan

SILS10460.10 SILS10460.25
SILNSILS10460.10EA 0 EUR
SILNSILS10460.10 SILNSILS10460.25
(3-Mercaptopropyl)triethoxysilan
(3-Mercaptopropyl)triethoxysilan
Formel: HSH₂CCH₂CH₂Si(OCH₂CH₃)₃
Molecular Weight: 238,42 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00053924
CAS-Nummer: 14814-09-6
EINECS: 238-883-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR