Methyldiphenylsilan
Lieferant: CHEMPUR SILANE
|
Achtung
|
Synonyme:
Diphenylmethylsilan
SILS08000.10
SILS08000.25
SILNSILS08000.10EA
0
EUR
SILNSILS08000.10
SILNSILS08000.25
Methyldiphenylsilan
Methyldiphenylsilan
Formel:
C₁₃H₁₄Si Molecular Weight: 198,34 g/mol Siedepunkt: 267 °C (1013 hPa) Dichte: 0,9975 g/cm³ (20 °C) |
MDL:
MFCD00008255 CAS-Nummer: 776-76-1 EINECS: 212-281-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...