Triisobutylsilan

Lieferant: CHEMPUR SILANE

SILS17925.5 SILS17925.25
SILNSILS17925.5EA 0 EUR
SILNSILS17925.5 SILNSILS17925.25
Triisobutylsilan
Triisobutylsilan
Formel: C₁₂H₂₈Si
Molecular Weight: 200,44 g/mol
MDL: MFCD00010340
CAS-Nummer: 6485-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR