Triethylsilylacetylen 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Triethylsilylethin
L20217.03
L20217.06
L20217.03EA
28.5
EUR
L20217.03
L20217.06
Triethylsilylacetylen 97%
Triethylsilylacetylen
Purity: 97%
CAS No.: 1777-03-3
Molecular Formula: HC≡CSi(CH2CH3)3
Formula Weight: 140.31
Flash Point: 17°(62°F)
Refractive Index: 1.4315
Boiling Point: 136-140°
Beilstein Reference: 1743814
UN No.: 1993
MDL No.: MFCD00075373
Formel:
C₈H₁₆Si Molecular Weight: 140.3 g/mol Siedepunkt: 136…140 °C Dichte: 0.783 Flammpunkt: 17 °C (62 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075373 CAS-Nummer: 1777-03-3 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...