Triethylsilylacetylen
Lieferant: CHEMPUR SILANE
Synonyme:
Triethylsilylethin
SILS17615.5
SILS17615.25
SILNSILS17615.5EA
0
EUR
SILNSILS17615.5
SILNSILS17615.25
Triethylsilylacetylen
Triethylsilylacetylen
Formel:
C₈H₁₆Si Molecular Weight: 140,3 g/mol |
MDL:
MFCD00075373 CAS-Nummer: 1777-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...