Tetrabutylsilan
Lieferant: CHEMPUR SILANE
SILS15275.5
SILS15275.25
SILNSILS15275.5EA
0
EUR
SILNSILS15275.5
SILNSILS15275.25
Tetrabutylsilan
Tetrabutylsilan
Formel:
C₁₆H₃₆Si Molecular Weight: 256.55 g/mol |
CAS-Nummer:
994-79-6 EINECS: 213-621-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...