Tetraethylsilan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 631-36-7
Molecular Formula: C8H20Si
Formula Weight: 144.33
Flash Point: 26°(78°F)
Refractive Index: 1.4260
Odour: Recognizable
Melting Point: -82°
Boiling Point: 153-155°
Beilstein Reference: 1732768
UN No.: 1993
MDL No.: MFCD00009019
Formel:
C₈H₂₀Si Siedepunkt: 153…155 °C Schmelzpunkt: –82 °C Dichte: 0,767 Flammpunkt: 26 °C (78 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009019 CAS-Nummer: 631-36-7 EINECS: 211-155-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...