Cyclopentyltrimethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Trimethoxy(cyclopentyl)silane

SILS05375.5 SILS05375.25
SILNSILS05375.5EA 0 EUR
SILNSILS05375.5 SILNSILS05375.25
Cyclopentyltrimethoxysilan
Cyclopentyltrimethoxysilan
Formel: C₈H₁₈O₃Si
Molecular Weight: 190.31 g/mol
Siedepunkt: 75 °C (10 mmHg)
Dichte: 0.99 g/cm³ (20 °C)
MDL: MFCD00236509
CAS-Nummer: 143487-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR