Tetraphenylsilan

Lieferant: CHEMPUR SILANE

SILS16375.5 SILS16375.25
SILNSILS16375.5EA 0 EUR
SILNSILS16375.5 SILNSILS16375.25
Tetraphenylsilan
Tetraphenylsilane
Formel: C₂₄H₂₀Si
MDL: MFCD00014069
CAS-Nummer: 1048-08-4
EINECS: 213-881-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR