Bismut(III)chlorid, wasserfrei ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Bismuttrichlorid

89485.06 89485.14
89485.06EA 130 EUR
89485.06 89485.14
Bismut(III)chlorid, wasserfrei ≥99,999% (Metall-Basis)
Bismut(III)chlorid

Purity: 99.999% CAS No.: 7787-60-2 Molecular Formula: BiCl3 Formula Weight: 315.34 Physical Form: Crystalline Density: 4.75 Appearance: White Melting Point: 230-232° Boiling Point: ca 430° subl. Merck Reference: 14,1262 UN No.: 3260 MDL No.: MFCD00003461

Formel: BiCl₃
Molecular Weight: 315,34 g/mol
Siedepunkt: ∼430 °C (Subl.)
Schmelzpunkt: 230…232 °C
Dichte: 4,75
Lagertemperatur: Raumtemperatur
MDL: MFCD00003461
CAS-Nummer: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8,III
Merck Index: 14,1262

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR