Antimon(III)iodid, ultra trocken ≥99,998% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Antimontriiodid

Unter Argon in Ampullen verpackt

35676.14 35676.03
35676.14EA 753 EUR
35676.14 35676.03
Antimon(III)iodid, ultra trocken ≥99,998% (Metall-Basis)
Antimon(III)iodid

Purity: 99.998% CAS No.: 7790-44-5 Molecular Formula: SbI3 Formula Weight: 502.46 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Moisture Sensitive Boiling Point: 420° Solubility: Decomposed by water to SbOI. Merck Reference: 14,710 UN No.: 3260 MDL No.: MFCD00016322

Formel: I₃Sb
Molecular Weight: 502,47 g/mol
Siedepunkt: 420 °C
Schmelzpunkt: 168 °C
Dichte: 4,676 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016322
CAS-Nummer: 7790-44-5
EINECS: 232-205-8
UN: 3260
ADR: 8,II
Merck Index: 14,710

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR