Arsen(III)iodid ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
41962.06
41962.14
41962.06EA
167
EUR
41962.06
41962.14
Arsen(III)iodid ≥99,999% (Metall-Basis)
Arsen(III)iodid
Purity: 99.999%
CAS No.: 7784-45-4
Molecular Formula: AsI3
Formula Weight: 455.64
Physical Form: -80 Mesh Powder
Density: 4.39
Melting Point: 140°
Boiling Point: 424°
Merck Reference: 14,803
UN No.: 1557
MDL No.: MFCD00014170
Formel:
AsI₃ Molecular Weight: 455,64 g/mol Siedepunkt: 424 °C Schmelzpunkt: 140 °C Dichte: 4,39 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014170 CAS-Nummer: 7784-45-4 EINECS: 232-068-4 UN: 1557 ADR: 6.1,II Merck Index: 14,803 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...