Bismut(III)iodid ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Bismuth triiodide

10656.04 10656.09 10656.18
10656.04EA 45.9 EUR
10656.04 10656.09 10656.18
Bismut(III)iodid ≥99,999% (Metall-Basis), Puratronic®
Bismut(III)iodid

Purity: 99.999% CAS No.: 7787-64-6 Molecular Formula: BiI3 Formula Weight: 589.69 Physical Form: Lump/Powder Density: 5.778 Appearance: Dark grey to black Odour: Acidic Melting Point: 408° Boiling Point: ca 500° Merck Reference: 14,1269 UN No.: 3260 MDL No.: MFCD00010894

Formel: BiI₃
Molecular Weight: 589,69 g/mol
Siedepunkt: ∼500 °C
Schmelzpunkt: 408 °C
Dichte: 5,778
Lagertemperatur: Raumtemperatur
MDL: MFCD00010894
CAS-Nummer: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8,III
Merck Index: 14,1269

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR