Antimon(III)oxid ≥99,6%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Antimontrioxid, Diantimontrioxid
11579.18
11579.36
11579.A3
11579.18EA
34.6
EUR
11579.18
11579.36
11579.A3
Antimon(III)oxid ≥99,6%
Antimon(III)oxid
Purity: 99.6% min
CAS No.: 1309-64-4
Molecular Formula: Sb2O3
Formula Weight: 291.50
Physical Form: 1.1-1.8 Micron Powder
Density: 5.67
Appearance: White
Odour: Odourless
Melting Point: 656°
Boiling Point: 1550° subl.
Solubility: Slightly soluble in water, Slowly dissolves in warm HCl or warm KOH solutions
Merck Reference: 14,711
MDL No.: MFCD00011214
Formel:
O₃Sb₂ Molecular Weight: 291,52 g/mol Siedepunkt: 1550 °C (Subl.) Schmelzpunkt: 656 °C Dichte: 5,67 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011214 CAS-Nummer: 1309-64-4 EINECS: 215-175-0 UN: 1549 ADR: 6.1,III Merck Index: 14,711 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...