Antimon(III)oxid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Diantimontrioxid
, Antimontrioxid
M-Clarity™ quality level = MQ200
SIAL230898-100GEA
47.2
EUR
SIAL230898-100G
SIAL230898-1KG
Antimon(III)oxid, Sigma-Aldrich®
Antimon(III)oxid
Formel:
O₃Sb₂ Molecular Weight: 291,52 g/mol Siedepunkt: 1425 °C (1013 hPa) Schmelzpunkt: 654 °C Dichte: 5,2 g/cm³ (20 °C) |
MDL:
MFCD00011214 CAS-Nummer: 1309-64-4 EINECS: 215-175-0 UN: 1549 ADR: 6.1,III Merck Index: 12,00752 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...