Arsen(III)oxid 99,95-100,05% ACS primärer Standard
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
33289.14
33289.22
33289.14EA
115
EUR
33289.14
33289.22
Arsen(III)oxid 99,95-100,05% ACS primärer Standard
Arsen(III)oxid
Purity: 99.95-100.05
CAS No.: 1327-53-3
Molecular Formula: As2O3
Formula Weight: 197.84
Physical Form: Powder
Density: 3.74
Melting Point: 312.3°
Boiling Point: 465° subl.
Merck Reference: 14,804
UN No.: 1561
MDL No.: MFCD00003433
Formel:
As₂O₃ Siedepunkt: 465 °C (Subl.) Schmelzpunkt: 312,3 °C Dichte: 3,74 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003433 CAS-Nummer: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1,II Merck Index: 14,804 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...