Arsen(III)oxid ≥99,99% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: di-Arsentrioxid, Arsenanhydrid

40370.09 40370.18 40370.30
40370.09EA 71.7 EUR
40370.09 40370.18 40370.30
Arsen(III)oxid ≥99,99% (Metall-Basis)
Arsen(III)oxid

Purity: 99.99% CAS No.: 1327-53-3 Molecular Formula: As2O3 Formula Weight: 197.84 Physical Form: -100 Mesh Powder Density: 3.74 Melting Point: 312.3° Boiling Point: 465° subl. Merck Reference: 14,804 UN No.: 1561 MDL No.: MFCD00003433

Formel: As₂O₃
Molecular Weight: 197,84 g/mol
Siedepunkt: 465 °C (Subl.)
Schmelzpunkt: 312,3 °C
Dichte: 3,74
Lagertemperatur: Raumtemperatur
MDL: MFCD00003433
CAS-Nummer: 1327-53-3
EINECS: 215-481-4
UN: 1561
ADR: 6.1,II
Merck Index: 14,804

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR