Bismut(III)oxid ≥99,99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dibismuttrioxid
17132.30
17132.18
17132.30EA
244
EUR
17132.30
17132.18
Bismut(III)oxid ≥99,99% (Metall-Basis)
Bismut(III)oxid
Purity: 99.99%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: -325 Mesh Powder
Density: 8.9
Melting Point: 817°
Boiling Point: ca 1890°
Merck Reference: 14,1273
MDL No.: MFCD00003462
Formel:
BiO₃ Molecular Weight: 465,96 g/mol Siedepunkt: ∼1890 °C Schmelzpunkt: 817 °C Dichte: 8,9 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003462 CAS-Nummer: 1304-76-3 EINECS: 215-134-7 Merck Index: 14,1273 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...