Triethylphosphit ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L00339.AE L00339.AP
L00339.AEEA 23.8 EUR
L00339.AE L00339.AP
Triethylphosphit ≥98%
Triethylphosphit

Purity: 98% CAS No.: 122-52-1 Molecular Formula: C6H15O3P Formula Weight: 166.16 Flash Point: 43°(109°F) Refractive Index: 1.4130 Odour: Unpleasant Melting Point: -112° Boiling Point: 155-157° Beilstein Reference: 956578 UN No.: 2323 MDL No.: MFCD00009084

Formel: C₆H₁₅O₃P
Siedepunkt: 155…157 °C
Schmelzpunkt: –112 °C
Dichte: 0,958
Flammpunkt: 43 °C (109 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009084
CAS-Nummer: 122-52-1
EINECS: 204-552-5
UN: 2323
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR