Antimon(V)sulfid, rein
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
di-Antimonpentasulfid, Antimonpentasulfid
330410010
330410500
330412500
ACRO330410010EA
198
EUR
ACRO330410010
ACRO330410500
ACRO330412500
Antimon(V)sulfid, rein
Antimon(V)sulfid
Formel:
S₅Sb₂ Molecular Weight: 403,85 g/mol Dichte: 4,12 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016321 CAS-Nummer: 1315-04-4 EINECS: 215-255-5 UN: 3179 ADR: 4.1,II Merck Index: 12,00738 |
Spezifikation Testergebnisse
Appearance | Yellow to orange-red Powder |
Assay | ≥54 % (Sb) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...