Arsen(III)sulfid ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Arsentrisulfid, di-Arsentrisulfid

41980.06 41980.14
41980.06EA 118 EUR
41980.06 41980.14
Arsen(III)sulfid ≥99,999% (Metall-Basis)
Arsen(III)sulfid

Purity: 99.999% CAS No.: 1303-33-9 Molecular Formula: As2S3 Formula Weight: 246.03 Physical Form: 12mm & Down Fused Lump Density: 3.43 Melting Point: 300° Boiling Point: 707° Merck Reference: 14,806 UN No.: 1557 MDL No.: MFCD00003435


For vacuum deposition

Formel: As₂S₃
Molecular Weight: 246,04 g/mol
Siedepunkt: 707 °C
Schmelzpunkt: 300 °C
Dichte: 3,43
Lagertemperatur: Raumtemperatur
MDL: MFCD00003435
CAS-Nummer: 1303-33-9
EINECS: 215-117-4
UN: 1557
ADR: 6.1,III
Merck Index: 14,806

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR