Bismut(III)sulfid ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dibismuttrisulfid
35834.18
35834.30
35834.18EA
166
EUR
35834.18
35834.30
Bismut(III)sulfid ≥99,9% (Metall-Basis)
Bismut(III)sulfid
Purity: 99.9%
CAS No.: 1345-07-9
Molecular Formula: Bi2S3
Formula Weight: 514.15
Physical Form: Chunks/Crystals
Density: 7.7
Appearance: Brown/black
MDL No.: MFCD00014204
Formel:
Bi₂S₃ Molecular Weight: 514,16 g/mol Schmelzpunkt: 763 °C Dichte: 7,7 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014204 CAS-Nummer: 1345-07-9 EINECS: 215-716-0 Merck Index: 12,01329 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...