Bismut(III)sulfid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
45463.30
45463.18
45463.30EA
1367
EUR
45463.30
45463.18
Bismut(III)sulfid ≥99,999% (Metall-Basis)
Bismut(III)sulfid
Purity: 99.999%
CAS No.: 1345-07-9
Molecular Formula: Bi2S3
Formula Weight: 514.15
Physical Form: Lump
Density: 7.7
Appearance: Gray
MDL No.: MFCD00014204
Formel:
Bi₂S₃ Schmelzpunkt: 763 °C Dichte: 7,7 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014204 CAS-Nummer: 1345-07-9 EINECS: 215-716-0 Merck Index: 12,01329 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...