(Methyl)triphenylphosphoniumbromid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
130079-500G
130079-100G
130079-25G
SIAL130079-500GEA
177
EUR
SIAL130079-500G
SIAL130079-100G
SIAL130079-25G
(Methyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(Methyl)triphenylphosphoniumbromid
Formel:
C₁₉H₁₈BrP Molecular Weight: 357.23 g/mol Schmelzpunkt: 232 Flammpunkt: >240 °C |
MDL:
MFCD00011804 CAS-Nummer: 1779-49-3 EINECS: 217-218-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...