Tetraethylammoniumtetrafluoroborat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Tetrylammonium tetrafluoroborate

A10211.06 A10211.18 A10211.22 A10211.30
A10211.06EA 23 EUR
A10211.06 A10211.18 A10211.22 A10211.30
Tetraethylammoniumtetrafluoroborat ≥99%
Tetraethylammoniumtetrafluoroborat

Purity: 99% CAS No.: 429-06-1 Molecular Formula: C8H20BF4N Formula Weight: 217.06 Melting Point: 370-380° dec. Beilstein Reference: 3917638 UN No.: 2811 MDL No.: MFCD00011827

Formel: C₈H₂₀BF₄N
Molecular Weight: 217.06 g/mol
Schmelzpunkt: 370…380 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011827
CAS-Nummer: 429-06-1
EINECS: 207-055-1
UN: 2811
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR