Tetraethylammoniumtetrafluoroborat, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Tetrylammonium tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ100

86618-25G 86618-5G
86618-25GEA 363 EUR
86618-25G 86618-5G
Tetraethylammoniumtetrafluoroborat, Supelco®
Tetraethylammoniumtetrafluoroborat
Formel: C₈H₂₀BF₄N
Molecular Weight: 217,06 g/mol
Schmelzpunkt: 365…368 °C
MDL: MFCD00011827
CAS-Nummer: 429-06-1
EINECS: 207-055-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2566d37a3d73a66aa93049f43a207e39ac2bbda7&vid=f63a12bf4834bebc4a30b3f2d97078f3edb33d0b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR