Cetrimoniumbromid, RonaCare®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Hexadecyltrimethylammoniumbromid, N-Cetyl-N,N,N-trimethylammoniumbromid, CTAB, N-Hexadecyl-N,N,N-trimethylammoniumbromid, Cetyltrimethylammoniumbromid
1.02343.1000
1.02343.9025
1.02343.0100
1.02343.1000EA
345
EUR
1.02343.1000
1.02343.9025
1.02343.0100
Cetrimoniumbromid, RonaCare®
Cetrimoniumbromid
Formel:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ Molecular Weight: 364,45 g/mol Schmelzpunkt: 237…243 °C Flammpunkt: 244 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011772 CAS-Nummer: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9,III Merck Index: 13,02034 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...