Tetrabutylammoniumbromid 99+%

Lieferant: Thermo Fisher Scientific
Achtung

185680250 185681000 185685000 185680500 185680025
185680250.EA 27.9 EUR
185680250. 185681000. 185685000. 185680500. 185680025.
Tetrabutylammoniumbromid 99+%
Tetrabutylammoniumbromid
Formel: C₁₆H₃₆BrN
Molecular Weight: 322.37 g/mol
Schmelzpunkt: 100…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011633
CAS-Nummer: 1643-19-2
EINECS: 216-699-2

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Water ≤1 %
HPLC ≥94 % (sum of tylosins A+B+C+D)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR