Tetraethylammoniumbromid 99+%

Lieferant: Thermo Scientific

Synonyme: Tetrylammoniumbromid

ACRO220715000EA 229 EUR
ACRO220715000 ACRO220710500 ACRO220712500 ACRO220710025
Tetraethylammoniumbromid 99+%
Tetraethylammoniumbromid
Formel: C₈H₂₀BrN
Molecular Weight: 210,16 g/mol
Schmelzpunkt: 285 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011825
CAS-Nummer: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Loss on drying ≤1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR