Tetraoctylammoniumbromid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Tetra-n-octylammonium bromide
352100050
352100250
352101000
ACRO352100050EA
42.9
EUR
ACRO352100050
ACRO352100250
ACRO352101000
Tetraoctylammoniumbromid 98%
Tetraoctylammoniumbromid
Formel:
C₃₂H₆₈BrN Molecular Weight: 546,8 g/mol Schmelzpunkt: 95…99 °C Flammpunkt: 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011863 CAS-Nummer: 14866-33-2 EINECS: 238-936-9 |
Spezifikation Testergebnisse
Appearance | White to cream Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 95 to 99 °C |
Titration Argentometric | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...