Tetraoctylammoniumbromid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Tetra-n-octylammonium bromide
A14929.09
A14929.18
A14929.30
A14929.09EA
56.3
EUR
A14929.09
A14929.18
A14929.30
Tetraoctylammoniumbromid ≥98%
Tetraoctylammoniumbromid
Purity: 98+%
CAS No.: 14866-33-2
Molecular Formula: [CH3(CH2)7]4NBr
Formula Weight: 546.81
Melting Point: 94-99°
Beilstein Reference: 4169176
Formel:
C₃₂H₆₈BrN Molecular Weight: 546,8 g/mol Schmelzpunkt: 94…99 °C Flammpunkt: 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011863 CAS-Nummer: 14866-33-2 EINECS: 238-936-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...