Tetrapropylammoniumbromid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
TPABr
M-Clarity™ Qualitätsstufe = MQ100
225568-100G
225568-500G
SIAL225568-100GEA
66.5
EUR
SIAL225568-100G
SIAL225568-500G
Tetrapropylammoniumbromid, Sigma-Aldrich®
Tetrapropylammoniumbromid
Formel:
C₁₂H₂₈BrN Molecular Weight: 266.27 g/mol Schmelzpunkt: 265…274 °C |
MDL:
MFCD00011840 CAS-Nummer: 1941-30-6 EINECS: 217-727-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...