Chlormequatchlorid, (max. 5% H₂O) 98% (Trockengewicht)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (2-Chlorethyl)trimethylammoniumchlorid, Chlorcholinchlorid, 2-Chlorethyltrimethylammoniumchlorid

A15630.06 A15630.14 A15630.22
A15630.06EA 18.3 EUR
A15630.06 A15630.14 A15630.22
Chlormequatchlorid, (max. 5% H₂O) 98% (Trockengewicht)
Chlormequatchlorid

Purity: 98% CAS No.: 999-81-5 Molecular Formula: C5H12ClN.HCl Formula Weight: 158.07 Odour: Amine-like Melting Point: ca 241° dec. Beilstein Reference: 3563994 Merck Reference: 14,2104 UN No.: 2811 MDL No.: MFCD00011869

Formel: ClCH₂CH₂N(CH₃)₃Cl
Molecular Weight: 158.07 g/mol
Schmelzpunkt: ∼ 241 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011869
CAS-Nummer: 999-81-5
EINECS: 213-666-4
UN: 2811
ADR: 6.1 III
Merck Index: 14,2104

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR