Acetylcholinchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Acetylcholiniumchlorid, (2-Acetoxyethyl)trimethylammoniumchlorid

L02168.14 L02168.22
L02168.14EA 45.9 EUR
L02168.14 L02168.22
Acetylcholinchlorid ≥98%
Acetylcholinchlorid

Purity: 98+% CAS No.: 60-31-1 Molecular Formula: CH3CO2CH2CH2N(CH3)3Cl Formula Weight: 181.66 Storage Temperature: 2°C to 8°C Melting Point: 149-152° Beilstein Reference: 3571875 Merck Reference: 14,87 MDL No.: MFCD00011698

Formel: H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
Molecular Weight: 181.66 g/mol
Schmelzpunkt: 149…152 °C
Dichte: 1.263 g/cm³ (20 °C)
MDL: MFCD00011698
CAS-Nummer: 60-31-1
EINECS: 200-468-8
Merck Index: 13,00089

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR