Acetylthiocholinchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acetylthiocholiniumchlorid, (2-Acetylthioethyl)trimethylammoniumchlorid

M-Clarity™ Qualitätsstufe = MQ200

A5626-1G A5626-5G
SIALA5626-1GEA 135 EUR
SIALA5626-1G SIALA5626-5G
Acetylthiocholinchlorid, Sigma-Aldrich®
Acetylthiocholinchlorid
Formel: H₃CC(S)O(CH₂)₂N(CH₃)₃Cl
Molecular Weight: 197,73 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00038727
CAS-Nummer: 6050-81-3
EINECS: 227-953-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR